Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECRG4 Antibody (OTI3B10), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272294APCCY7
Description
ECRG4 Monoclonal specifically detects ECRG4 in Human samples. It is validated for Flow Cytometry.Specifications
ECRG4 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
C2orf40 | |
Full length recombinant protein of human ECRG4 (NP_115787) produced in HEK293T cells. | |
0.1 mL | |
84417.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI3B10 | |
Flow Cytometry | |
chromosome 2 open reading frame 40, ECRG4augurin, esophageal cancer related gene 4 protein, Esophageal cancer-related gene 4 protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction