Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECRG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572717
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572717 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572717 Supplier Invitrogen™ Supplier No. PA572717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECRG4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Probable hormone that may attenuate cell proliferation and induce senescence of oligodendrocyte and neural precursor cells in the central nervous system. ECRG4-induced senescence is characterized by G1 arrest, RB1 dephosphorylation and accelerated CCND1 and CCND3 proteasomal degradation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECRG4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene C2orf40
Gene Accession No. Q9H1Z8
Gene Alias Augurin; C2orf40; chromosome 2 open reading frame 40; ECRG4; esophageal cancer related gene 4 protein; esophageal cancer-related gene 4 protein; UNQ761/PRO1508
Gene Symbols C2orf40
Host Species Rabbit
Immunogen ECRG4 antibody for a 20 amino acid synthetic peptide near the carboxy terminus of human ECRG4. The immunogen is within the last 50 amino acids of ECRG4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84417
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.