Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECRG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104506
Description
Antibody detects endogenous levels of total ECRG4.
Probable hormone that may attenuate cell proliferation and induce senescence of oligodendrocyte and neural precursor cells in the central nervous system. ECRG4-induced senescence is characterized by G1 arrest, RB1 dephosphorylation and accelerated CCND1 and CCND3 proteasomal degradation. [UniProt].
Specifications
ECRG4 | |
Polyclonal | |
Unconjugated | |
C2orf40 | |
Augurin; C2orf40; chromosome 2 open reading frame 40; ECRG4; esophageal cancer related gene 4 protein; esophageal cancer-related gene 4 protein; UNQ761/PRO1508 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84417 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9H1Z8 | |
C2orf40 | |
A synthesized peptide derived from human ECRG4, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction