Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECS Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277834
Encompass
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7834 1 mg
50-227-7833 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-227-7834 Supplier Leinco Technologies Inc Supplier No. 188301MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ECS Polyclonal antibody specifically detects ECS in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ECS
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias DDDDK; ECS; ECS Cleavage Site
Host Species Goat
Immunogen DDDDK conjugated to KLH.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.