Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECSCR Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA538003
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is primarily found in endothelial cells and blood vessels, where it is involved in cell shape changes and EGF-induced cell migration. It can enhance the activation of vascular endothelial growth factor receptor-2/kinase insert domain receptor and also promote the proteolysis of internalized kinase insert domain receptor. This gene may play a role in angiogenesis-related diseases. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jun 2014].
Specifications
| ECSCR | |
| Polyclonal | |
| Unconjugated | |
| ECSCR | |
| 1110006O17Rik; apoptosis regulator through modulating IAP expression; ARIA; Ecscr; ECSM2; endothelial cell surface expressed chemotaxis and apoptosis regulator; endothelial cell-specific chemotaxis regulator; Endothelial cell-specific molecule 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 641700 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q19T08 | |
| ECSCR | |
| Peptide with sequence C-NNGKQSLSAEKVL, from the C-terminus of ECSM2 (aa 193-205). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction