Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECSCR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584594 Supplier Invitrogen™ Supplier No. PA584594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQPTMTQTSS SQGGLGGLSL TTEPVSSNPG YIPSSEANRP SHLSSTGT Highest antigen sequence indentity to the following orthologs: Rat - 37%, Mouse - 40%.

The protein encoded by this gene is primarily found in endothelial cells and blood vessels, where it is involved in cell shape changes and EGF-induced cell migration. It can enhance the activation of vascular endothelial growth factor receptor-2/kinase insert domain receptor and also promote the proteolysis of internalized kinase insert domain receptor. This gene may play a role in angiogenesis-related diseases. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jun 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECSCR
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ECSCR
Gene Accession No. Q19T08
Gene Alias 1110006O17Rik; apoptosis regulator through modulating IAP expression; ARIA; Ecscr; ECSM2; endothelial cell surface expressed chemotaxis and apoptosis regulator; endothelial cell-specific chemotaxis regulator; Endothelial cell-specific molecule 2
Gene Symbols ECSCR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ECSCR. Recombinant protein control fragment (Product #RP-108488).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.