Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECSIT Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300303MFV500
Description
ECSIT Polyclonal antibody specifically detects ECSIT in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
ECSIT | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved | |
Produced in rabbits immunized with purified, recombinant Human ECSIT (Uniprot#: Q9BQ95-1; Glu246-Ser431) | |
0.1 mL | |
Immunology, Innate Immunity, Signal Transduction, Wnt Signaling Pathway | |
51295 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction