Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECSIT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287320
Description
ECSIT Polyclonal specifically detects ECSIT in Mouse samples. It is validated for Western Blot.Specifications
ECSIT | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
ECSIT homolog (Drosophila), evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial, likely ortholog of mouse signaling intermediate in Toll pathway evolutionarilyconserved, Protein SITPEC, signaling intermediate in Toll pathway evolutionarily conserved ortholog, SITPECsignaling intermediate in Toll pathway, evolutionarily conserved | |
The immunogen is a synthetic peptide corresponding to a region of Mouse ECSIT | |
100 μL | |
Immunology, Innate Immunity, Signal Transduction, Wnt Signaling Pathway | |
51295 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction