Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECSIT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560183 Supplier Invitrogen™ Supplier No. PA560183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HMEPDLSARV TIYQVPLPKD STGAADPPQP HIVGIQSPDQ QAALARHNPA RPVFVEGPFS LWLRNKCVYY HILRADLLPP EEREVEETPE EWNLYYPM Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Activation of NF-kappa-B as a result of Toll-like receptor (TLR) and IL-1 receptor signaling is a major component of innate immune responses. Signals from these receptors are relayed by a number of adapter molecules such as TRIF, TIRAP, and MyD88 to kinases such as IRAK and other intermediates such as TNF receptor associated factor (TRAF)-6. ECSIT (evolutionarily conserved signaling intermediate in Toll pathways) was initially identified as a cytoplasmic protein interacting specifically with TNF receptor associated factor (TRAF)-6 in the TLR pathway. Recently however, ECSIT has also been shown to be required for bone morphogenetic protein (Bmp) signaling and mesoderm formation during mouse embryogenesis, indicating the possibility of cross-talk between the TLR/IL-B and Bmp signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECSIT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ECSIT
Gene Accession No. Q5XIC2, Q9BQ95, Q9QZH6
Gene Alias Ecsit; ECSIT homolog; ECSIT homolog (Drosophila); ECSIT signalling integrator; evolutionarily conserved signaling intermediate in Toll pathway, mitochondrial; likely ortholog of mouse signaling intermediate in Toll pathway evolutionarily conserved; Protein SITPEC; signaling intermediate in Toll pathway-evolutionarily conserved; Sitpec
Gene Symbols ECSIT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ECSIT. Recombinant protein control fragment (Product #RP-99074).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26940, 300447, 51295
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.