Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECT2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128729G
Description
ECT2 Polyclonal antibody specifically detects ECT2 in Human, Mouse samples. It is validated for Western BlotSpecifications
ECT2 | |
Polyclonal | |
Western Blot | |
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2 | |
The immunogen recognized by this antibody maps to a region between residue 833 and 883 of human epithelial cell transforming sequence 2 using the numbering given in entry NP_060568.3 (GeneID 1894). | |
0.1 mL | |
Phospho Specific | |
1894 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction