Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECT2 Antibody (OTI2D9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270613FR
Description
ECT2 Monoclonal specifically detects ECT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ECT2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ECT2 | |
Recombinant protein fragment corresponding to amino acids 175-438 of human ECT2 (NP_060568) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D9 | |
Western Blot, Immunohistochemistry | |
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2 | |
Mouse | |
Affinity Purified | |
Phospho Specific | |
1894.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction