Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECT2 Monoclonal Antibody (1691CT516.3.3)
GREENER_CHOICE

Catalog No. PIMA537721
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537721 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537721 Supplier Invitrogen™ Supplier No. MA537721
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ECT2 Monoclonal Antibody for Western Blot

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECT2
Applications Western Blot
Classification Monoclonal
Clone 1691CT516.3.3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ECT2
Gene Accession No. Q9H8V3
Gene Alias AI528536; ARHGEF31; Ect2; ect2 oncogene; ect2 protein; epithelial cell transforming 2; epithelial cell transforming sequence 2 oncogene; epithelial cell transforming sequence 2 oncogene protein; epithelial cell-transforming sequence 2 oncogene; FLJ10461; LOW QUALITY PROTEIN: protein ECT2; MGC138291; mKIAA4037; Protein ECT2; Protein ECT2-like protein
Gene Symbols ECT2
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 200-460 amino acids from human ECT2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1894
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.