Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECT2 Monoclonal Antibody (OTI2D9)
GREENER_CHOICE

Catalog No. PIMA526870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526870 Supplier Invitrogen™ Supplier No. MA526870
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ECT2 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ECT2
Gene Accession No. Q9H8V3
Gene Alias AI528536; ARHGEF31; Ect2; ect2 oncogene; ect2 protein; epithelial cell transforming 2; epithelial cell transforming sequence 2 oncogene; epithelial cell transforming sequence 2 oncogene protein; epithelial cell-transforming sequence 2 oncogene; FLJ10461; LOW QUALITY PROTEIN: protein ECT2; MGC138291; mKIAA4037; Protein ECT2; Protein ECT2-like protein
Gene Symbols ECT2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 175-438 of ECT2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1894
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.