Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518166
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518166 Supplier Invitrogen™ Supplier No. PA518166
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECT2
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ECT2
Gene Accession No. Q9H8V3
Gene Alias AI528536; ARHGEF31; Ect2; ect2 oncogene; ect2 protein; epithelial cell transforming 2; epithelial cell transforming sequence 2 oncogene; epithelial cell transforming sequence 2 oncogene protein; epithelial cell-transforming sequence 2 oncogene; FLJ10461; LOW QUALITY PROTEIN: protein ECT2; MGC138291; mKIAA4037; Protein ECT2; Protein ECT2-like protein
Gene Symbols ECT2
Host Species Goat
Immunogen Synthetic peptide sequence (AENSVLTSTTGRT) corresponding to the N-terminus amino acids of ECT2 (aa 2-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1894
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.