Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECT2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548246
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548246 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548246 Supplier Invitrogen™ Supplier No. PA548246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ECT2L Polyclonal Antibody for Western Blot, IHC (P)

May act as a guanine nucleotide exchange factor (GEF).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECT2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ECT2L
Gene Accession No. Q008S8
Gene Alias ARHGEF32; C6orf91; dJ509I19.2; dJ509I19.3; dJ509I19.5; ECT2L; ECT2-like; epithelial cell transforming 2 like; epithelial cell transforming sequence 2 oncogene-like; epithelial cell-transforming sequence 2 oncogene-like; F-box protein 49; FBXO49; LFDH; lung specific F-box and DH domain containing protein; lung-specific F-box and DH domain-containing protein; Putative guanine nucleotide exchange factor LFDH
Gene Symbols ECT2L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 725-753 amino acids from the C-terminal region of human ECT2L.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345930
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.