Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eculizumab Monoclonal Antibody (4F6), GenScript™

Catalog No. 502530049
Encompass
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-0049 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-253-0049 Supplier Genscript Corporation Supplier No. A0211540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. The lyophilized product remains stable up to 1 year at -20 °C from date of receipt. Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below. Avoid repeated freezing and thawing cycles.

Eculizumab Monoclonal antibody specifically detects Eculizumab in Chemical samples. It is validated for ELISA

Specifications

Antigen Eculizumab
Applications ELISA
Classification Monoclonal
Clone 4F6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.2
Gene Alias 5G1.1; h5G1.1HuG2-G4; Soliris
Host Species Mouse
Immunogen Eculizumab
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.