Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDA/Ectodysplasin Antibody (MM0254-3T2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | EDA/Ectodysplasin |
|---|---|
| Clone | MM0254-3T2 |
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EDA/Ectodysplasin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_001005609 | |
| 1896 | |
| IgG1 | |
| Protein G purified |
| MM0254-3T2 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| ectodysplasin A, ED1 | |
| EDA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title