Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDA/Ectodysplasin Antibody (Renzo-2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259772AF350
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
EDA/Ectodysplasin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
EDA | |
Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391). | |
0.1 mL | |
1896.0 | |
Human, Mouse | |
IgG1 |
Flow Cytometry, ELISA | |
Renzo-2 | |
Flow Cytometry, ELISA | |
ectodysplasin A, ED1 | |
Mouse | |
Protein G purified | |
Primary | |
Recognizes human and mouse EDA-A1 and EDA-A2. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction