Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDA/Ectodysplasin Antibody (Renzo-2), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259772PCP0.1ML
Description
EDA/Ectodysplasin Monoclonal specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
EDA/Ectodysplasin | |
Monoclonal | |
PerCP | |
PBS | |
EDA | |
Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391). | |
1896 | |
Human, Mouse | |
Purified |
Flow Cytometry | |
Renzo-2 | |
Flow Cytometry | |
ectodysplasin A, ED1 | |
Mouse | |
0.1 mL | |
Recognizes human and mouse EDA-A1 and EDA-A2. | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction