Learn More
Invitrogen™ EDA Polyclonal Antibody

Description
Immunogen sequence: ELRSELRRERG AESRLGGSGT PGTSGTLSSL GGLDPDSPIT SHLGQPSPKQ QPLEPGEAAL HS Highest antigen sequence identity to the following orthologs - mouse 78%, rat 78%.
Specifications
Specifications
| Antigen | EDA |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EDA |
| Gene Accession No. | Q92838 |
| Gene Alias | ECTD1; ectodermal dysplasia 1, anhidrotic; ectodermal dysplasia protein; Ectodysplasin; Ectodysplasin 1, isoform A2; ectodysplasin A; ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; Ed1; ED1-A1; ED1-A2; EDA; EDA 1; EDA protein; EDA protein homolog; EDA1; EDA2; Eda-A1; EDA-A2; HED; HED1; nackt; nkt; ODT1; oligodontia 1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; Tabby protein; TNLG7C; tumor necrosis factor ligand 7C; XHED; XLHED; X-linked anhidroitic ectodermal dysplasia protein |
| Gene Symbols | EDA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.