Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565199 Supplier Invitrogen™ Supplier No. PA565199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YYINFTDFASY EVVVDEKPFL QCTRSIETGK TNYNTCYTAG VCLLKARQKI AVKMVHADIS INMSKHTTFF GAIRLGEAPA S Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDA
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EDA
Gene Accession No. Q92838
Gene Alias ECTD1; ectodermal dysplasia 1, anhidrotic; ectodermal dysplasia protein; Ectodysplasin; Ectodysplasin 1, isoform A2; ectodysplasin A; ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; Ed1; ED1-A1; ED1-A2; EDA; EDA 1; EDA protein; EDA protein homolog; EDA1; EDA2; Eda-A1; EDA-A2; HED; HED1; nackt; nkt; ODT1; oligodontia 1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; Tabby protein; TNLG7C; tumor necrosis factor ligand 7C; XHED; XLHED; X-linked anhidroitic ectodermal dysplasia protein
Gene Symbols EDA
Host Species Rabbit
Immunogen Recombinant Human EDA. Recombinant protein control fragment (Product #RP-105942).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1896
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.