Learn More
Invitrogen™ EDA2R Polyclonal Antibody

Description
The antibody detects endogenous levels of total EDA2R protein.
Specifications
Specifications
| Antigen | EDA2R |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | EDA2R |
| Gene Accession No. | Q9HAV5 |
| Gene Alias | 9430060M22Rik; ectodysplasin A2 isoform receptor; ectodysplasin A2 receptor; EDA2R; EDA-A2 receptor; EDAA2R; EDA-A2R; RGD1564025; Tnfrsf27; Tumor necrosis factor receptor superfamily member 27; tumor necrosis factor receptor superfamily member XEDAR; UNQ2448/PRO5727/PRO34080; X linked ectodysplasin receptor; Xedar; X-linked ectodysplasin-A2 receptor |
| Gene Symbols | EDA2R |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.