Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDA2R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987739
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7739 Supplier Bioss Supplier No. BS7111R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

X-linked ectodysplasin-A2 receptor (XEDAR) is a recently isolated member of the tumor necrosis factor receptor family that is highly expressed during embryonic development and binds to ectodysplatin-A2 (EDA-A2). Two predominantly expressed isoforms, XEDAR-s and XEDAR-L, differ by only a 21-amino region at the juxtamembrane region of the cytoplasmic domain. Neither isoform possesses a death domain and both have been shown to act mainly through TRAF3 and TRAF6 to activate the NF-kappa-B and JNK pathways. Cells transfected with XEDAR and treated with EDA-A2 cause the assembly of a secondary complex containing FADD, caspase-8 and caspase-10, leading to the activation caspase-8 and caspase-3, and finally apoptosis. The EDA-A2-induced apoptosis is dependent on caspase-9 activation, as various pharmacological and genetic inhibitors of caspase-8 blocked apoptosis following EDA-A2 treatment.

Specifications

Antigen EDA2R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EDA2R
Gene Accession No. Q9HAV5
Gene Alias 9430060M22Rik; ectodysplasin A2 isoform receptor; ectodysplasin A2 receptor; EDA2R; EDA-A2 receptor; EDAA2R; EDA-A2R; Tnfrsf27; Tumor necrosis factor receptor superfamily member 27; tumor necrosis factor receptor superfamily member XEDAR; UNQ2448/PRO5727/PRO34080; X linked ectodysplasin receptor; XEDAR; X-linked ectodysplasin-A2 receptor
Gene Symbols EDA2R
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EDA2R/TNFRSF27.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60401
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.