Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDAR Antibody (19E8) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242695

Catalog No. NBP242695


Only null left
Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

EDAR Monoclonal specifically detects EDAR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

EDAR
Monoclonal
1 mg/mL
Western Blot, Flow Cytometry, ELISA
NP_071731
EDAR
Recombinant human EDAR (27-448aa) purified from E.coli.
100 μL
Apoptosis
10913
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, Flow Cytometry, ELISA
1900000000
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only