Learn More
Invitrogen™ EDAR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596290
Description
Immunogen sequence: EYSNCGENEY YNQTTGLCQE CPPCGPGEEP YLSCGYGTKD EDYGCVPCPA EKFSKGGYQI CRRHKDCEGF FRATVLTPGD MENDAECGPC LPGYYMLENR PRNIYGMVCY SCLLAPPNTK ECVGATSGAS ANFPGTSGSS TLSPFQHAHK ELSGQGHLAT A; Positive Samples: 293T, LO2, SW480; Cellular Location: Membrane, Single-pass type I membrane protein.
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
Specifications
EDAR | |
Polyclonal | |
Unconjugated | |
Edar | |
Anhidrotic ectodysplasin receptor 1; DL; Downless; downless homolog; downless, mouse, homolog of; ECTD10A; ECTD10B; ectodermal dysplasia receptor; ectodysplasin 1, anhidrotic receptor; ectodysplasin A receptor; ectodysplasin-A receptor; ED1R; ED3; ED5; EDA1R; EDA3; EDA-A1 receptor; EDA-A1R; Edar; HRM1; RGD1561714; Tumor necrosis factor receptor superfamily member EDAR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10913 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.88 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UNE0 | |
Edar | |
Recombinant fusion protein containing a sequence corresponding to amino acids 27-187 of human EDAR (NP_0717311). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.