Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDAR Recombinant Mouse Monoclonal Antibody (6H2), Invitrogen™

Catalog No. FEMA550237
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550237 Supplier Thermo Scientific Supplier No. MA550237

Mouse Recombinant Monoclonal Antibody

This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDAR
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 6H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300, pH 7.4
Gene Edar
Gene Accession No. Q9UNE0
Gene Alias Anhidrotic ectodysplasin receptor 1, DL, Downless, downless homolog, downless, mouse, homolog of, ECTD10A, ECTD10B, ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, ectodysplasin-A receptor, ED1R, ED3, ED5, EDA1R, EDA3, EDA-A1 receptor, EDA-A1R, Edar, HRM1, RGD1561714, Tumor necrosis factor receptor superfamily member EDAR
Gene Symbols Edar
Host Species Mouse
Immunogen Recombinant Human EDAR protein
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10913
Target Species Human
Content And Storage -20°C or -80°C if preferred
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.