Learn More
Invitrogen™ EDARADD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120410
Description
Positive test controls include: A-431, MCF7. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MGLRTTKQMG RGTKAPGHQE DHMVKEPVED TDPSTLSFNM SDKYPIQDTE LPKAEECDTI TLNCPRNSDM KNQGEENGFP DSTGDPLPEI SKDNSCKENC TCSSCLLRAP TISDLLNDQD LLDVIRIKLD PCHPTVKNWR NFASKWGMSY DELCFLEQRP QSPTLEFLLR NSQRTVGQLM ELCRLYHRAD VEKVLRRWVD EEWPKRERGD PSRHF.
Specifications
| EDARADD | |
| Polyclonal | |
| Unconjugated | |
| EDARADD | |
| crinkled homolog; ECTD11A; ECTD11B; ectodysplasia A receptor associated death domain; ectodysplasin A receptor associated adapter protein; ectodysplasin-A receptor-associated adapter protein; ED3; EDA3; EDAR associated death domain; EDARADD; EDAR-associated death domain protein; Protein crinkled homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 128178 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8WWZ3 | |
| EDARADD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human EDARADD (NP_665860.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.