Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDB-FN Chimeric Recombinant Rabbit Monoclonal Antibody (A2)
GREENER_CHOICE

Catalog No. PIMA548023
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548023 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548023 Supplier Invitrogen™ Supplier No. MA548023
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody bind to human Extra-Domain containing fibronectin.

EDB-FN is a variant of the fibronectin protein that contains an extra domain, known as the extra domain B. This extra domain is not present in the normal fibronectin protein and has been found to be expressed in certain tissues during embryonic development and in tumors. The extra domain has been implicated in cell adhesion, migration, and invasion, and has been shown to play a role in the progression and metastasis of certain cancers. Due to its unique expression pattern and functional properties, EDB-FN has been investigated as a potential target for cancer therapy and imaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDB-FN Chimeric
Applications ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene FN1
Gene Accession No. P02751
Gene Alias extra domain B; extra domain B of fibronectin
Gene Symbols FN1
Host Species Rabbit
Immunogen 2D-PAGE gel isolated human Extra-Domain containing fibronectin from lysate of human melanoma COLO-38 cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.