Learn More
Invitrogen™ EDB-FN Chimeric Recombinant Rabbit Monoclonal Antibody (G4)

Description
Specificity: This antibody bind to human Extra-Domain containing fibronectin.
Specifications
Specifications
| Antigen | EDB-FN Chimeric |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | G4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | FN1 |
| Gene Accession No. | P02751 |
| Gene Alias | extra domain B; extra domain B of fibronectin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.