Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522395
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522395 Supplier Invitrogen™ Supplier No. PA522395
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, NIH-3T3, C2C12. Predicted reactivity: Mouse (95%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is important in mRNA degradation. The encoded protein is a component of a decapping complex that promotes efficient removal of the monomethylguanosine (m7G) cap from mRNAs, as part of the 5' to 3' mRNA decay pathway. Mutations in this gene have been identified in human patients with an autosomal recessive form of intellectual disability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Edc3
Gene Accession No. Q8K2D3, Q96F86
Gene Alias AA517853; EDC3; enhancer of mRNA decapping 3; enhancer of mRNA decapping 3 homolog; enhancer of mRNA decapping 3 homolog (S. cerevisiae); enhancer of mRNA-decapping protein 3; hYjeF_N2; hYjeF_N2-15q23; hypothetical protein FLJ21128; hypothetical protein LOC504054; im:7151288; Lsm16; LSM16 homolog; LSM16 homolog (EDC3, S. cerevisiae); LSM16 protein homolog; MRT50; PP844; RGD1306898; wu:fb82d07; Yjdc; yjeF domain containing; yjeF domain-containing protein 1; YjeF N-terminal domain-containing protein 2; yjeF_N2; YJEFN2; zgc:112006
Gene Symbols Edc3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Edc3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353190, 80153
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.