Learn More
Invitrogen™ EDC3 Polyclonal Antibody

Description
Immunogen sequence: IDTYERRSGT RSRGIPNERP TRYRHDENIL ESEPIVYRRI IVPHNVSKEF CTDSGLVVPS ISYELHKKLL SVAEKHGLTL ERRLE Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.
Specifications
Specifications
| Antigen | EDC3 |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Edc3 |
| Gene Accession No. | Q96F86 |
| Gene Alias | AA517853; EDC3; enhancer of mRNA decapping 3; enhancer of mRNA decapping 3 homolog; enhancer of mRNA decapping 3 homolog (S. cerevisiae); enhancer of mRNA-decapping protein 3; hYjeF_N2; hYjeF_N2-15q23; hypothetical protein FLJ21128; hypothetical protein LOC504054; im:7151288; Lsm16; LSM16 homolog; LSM16 homolog (EDC3, S. cerevisiae); LSM16 protein homolog; MRT50; PP844; RGD1306898; wu:fb82d07; Yjdc; yjeF domain containing; yjeF domain-containing protein 1; YjeF N-terminal domain-containing protein 2; yjeF_N2; YJEFN2; zgc:112006 |
| Gene Symbols | Edc3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.