Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559461 Supplier Invitrogen™ Supplier No. PA559461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VERALETRHE QEQRRLERAL AEGQQRGGQL QEQLTQQLSQ ALSSAVAGRL ERSIRDEIKK TVPPCVSRSL EPMAGQLSNS VATKLTAVEG SMKENISKLL KSKNLTDA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

In the process of mRNA degradation, seems to play a role in mRNA decapping. Component of a complex containing DCP2 and DCP1A which functions in decapping of ARE-containing mRNAs. Promotes complex formation between DCP1A and DCP2. Enhances the catalytic activity of DCP2 (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDC4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EDC4
Gene Accession No. Q6P2E9
Gene Alias Autoantigen Ge-1; autoantigen RCD8; autoantigen RCD-8; BC022641; Edc4; enhancer of mRNA decapping 4; enhancer of mRNA-decapping protein 4; fe26b05; GE1; Ge-1; HEDL5; HEDLS; Human enhancer of decapping large subunit; LRRG00115; Nrn1l; RCD8; RCD-8; RGD1309136; si:ch211-237c7.1; si:dkey-16n13.6; wu:fe26b05
Gene Symbols EDC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EDC4. Recombinant protein control fragment (Product #RP-98599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23644
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.