Learn More
Invitrogen™ EDC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590452
Description
Immunogen sequence: GDGDRHNTPS LLEAALTQEA STPDSQVWPT APDITRETCS TLAESPRNGL QEKHKSLAFH RPPYHLLQQR DSQDASAEQS DHDDEVASLA SASGGFGTKV PAPRLPAKDW KTKGSPRTSP KLKRKSKKDD GDAAMGSRLT EHQVAEPPED WPALIWQQQR ELAELRHSQE ELLQRLCTQL EGLQSTVTGH VERALETRHE QEQRRLERAL AEGQQRGGQL QEQLTQQLSQ ALSSAVAGRL ERSIRDEIKK TVPPCVSRSL EPMAGQLSNS VATKLTAVEG SMKENISKLL KSKNLTDAIA R; Positive Samples: Jurkat, Mouse liver; Cellular Location: Cytoplasm, Nucleus, P-body.
In the process of mRNA degradation, seems to play a role in mRNA decapping. Component of a complex containing DCP2 and DCP1A which functions in decapping of ARE-containing mRNAs. Promotes complex formation between DCP1A and DCP2. Enhances the catalytic activity of DCP2 (in vitro).
Specifications
EDC4 | |
Polyclonal | |
Unconjugated | |
EDC4 | |
Autoantigen Ge-1; autoantigen RCD8; autoantigen RCD-8; BC022641; Edc4; enhancer of mRNA decapping 4; enhancer of mRNA-decapping protein 4; fe26b05; GE1; Ge-1; HEDL5; HEDLS; Human enhancer of decapping large subunit; LRRG00115; Nrn1l; RCD8; RCD-8; RGD1309136; si:ch211-237c7.1; si:dkey-16n13.6; wu:fe26b05 | |
Rabbit | |
Affinity chromatography | |
RUO | |
234699, 23644 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.67 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q3UJB9, Q6P2E9 | |
EDC4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 800-1100 of human EDC4 (NP_0551443). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.