Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDDM3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551559 Supplier Invitrogen™ Supplier No. PA551559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVSWREFMKQ HYLSPSREFR EYKCDVLMRE NEALKDKSSH MFIYISWYKI EHICTSDNWM DRFRNAYVWV QNPLKVLKCH QENSKNSYTE SRSFNYIEFH CSMDGYVDSI EDLKMVEPIG N Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 58%.

Testicular sperm are morphologically differentiated but are not progressively motile nor able to fertilize an egg. Post-testicular maturation requires exposure of spermatozoa to the microenvironment of the epididymal lumen. Spermatozoa undergo extensive changes in the epididymis, including enzymatic modifications, loss of pre-existing components and addition of new glycoproteins from epididymal secretions. These modifying proteins and enzymes are synthesized by epithelial cells lining the epididymal duct and secreted apically into the lumen, where they come into contact with, and may be absorbed onto, the sperm membranes. The proteins encoded by the genes in this cluster are synthesized and secreted by epididymal epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDDM3B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EDDM3B
Gene Accession No. P56851
Gene Alias EDDM3B; EP3B; epididymal protein 3B; epididymal secretory protein E3 beta; epididymal secretory protein E3-beta; FAM12B; family with sequence similarity 12, member B (epididymal); HE3B; HE3-BETA; human epididymis-specific 3 beta; human epididymis-specific protein 3-beta; RAM2; ribonuclease A M2; UNQ6412/PRO21187
Gene Symbols EDDM3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EDDM3B. Recombinant protein control fragment (Product #RP-88881).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.