Learn More
Description
Specifications
Specifications
| Antigen | EDEM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q92611 |
| Gene Alias | EDEMKIAA0212ER degradation-enhancing alpha-mannosidase-like 1, ER degradation enhancer, mannosidase alpha-like 1, FLJ51559, FLJ51560 |
| Gene Symbols | EDEM1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
