Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDEM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120535 Supplier Invitrogen™ Supplier No. PA5120535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A375, HepG2. Immunogen sequence: SIEKISKVEC GFATIKDLRD HKLDNRMESF FLAETVKYLY LLFDPTNFIH NNGSTFDAVI TPYGECILGA GGYIFNTEAH PIDPAALHCC QRLKEEQWEV EDLMREFYSL KRSRSKFQKN TVSSGPWEPP ARPGTLFSPE NHDQARERKP AKQKVPLLSC PSQPFTSKLA LLGQVFLDSS.

In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). EDEM2 belongs to a family of proteins involved in ERAD of glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDEM2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EDEM2
Gene Accession No. Q9BV94
Gene Alias 9530090G24Rik; AI327354; bA4204.1; C20orf31; C20orf49; Edem2; ER degradation enhancer, mannosidase alpha-like 2; ER degradation enhancing alpha-mannosidase like protein 2; ER degradation-enhancing alpha-mannosidase-like 2; ER degradation-enhancing alpha-mannosidase-like protein 2; ER degradation-enhancing-mannosidase-like protein 2; RGD1303074; UNQ573/PRO1135
Gene Symbols EDEM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 399-578 of human EDEM2 (NP_060687.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108687, 55741
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.