Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDEM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555253
Description
Immunogen sequence: FRSGAKPPLR ARDFMATNPE HLEILKKMGV SLIHLKDGRV QLVQHAIQAA SSIDAEDGLR FMQEMIELSS QQQKEQQLPP RAVQIVSHPF FGRVVLTAGP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.
Specifications
| EDEM3 | |
| Polyclonal | |
| Unconjugated | |
| EDEM3 | |
| 2310050N11Rik; Alpha-1,2-mannosidase EDEM3; C1orf22; EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation enhancing alpha-mannosidase like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3; ER degradation-enhancing -mannosidase-like protein 3; RGD1561496 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZQ6 | |
| EDEM3 | |
| Recombinant protein corresponding to Human EDEM3. Recombinant protein control fragment (Product #RP-94076). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction