Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDEM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565009
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565009 Supplier Invitrogen™ Supplier No. PA565009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSQEVDLVDQE SSEENSLNSH PESLSLADMD NAASISPSEQ TSNPTENHET TNLNGECTDL DNQLQEQSET EEDSNPNVSW GKKVQPIDSI LADWNEDIEA FEMMEKDE Highest antigen sequence identity to the following orthologs - mouse 72%, rat 26%.

Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDEM3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EDEM3
Gene Accession No. Q9BZQ6
Gene Alias 2310050N11Rik; Alpha-1,2-mannosidase EDEM3; C1orf22; EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation enhancing alpha-mannosidase like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3; ER degradation-enhancing -mannosidase-like protein 3; RGD1561496
Gene Symbols EDEM3
Host Species Rabbit
Immunogen Recombinant Human EDEM3. Recombinant protein control fragment (Product #RP-105774).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.