Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium, Dihydrate, BiotechGrade, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18602380 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-380 6 x 500 g Poly Bottle
18-602-379 500 g Poly Bottle
18-602-381 2.5 kg Poly Bottle
18-602-382 4 x 2.5 kg Poly Bottle
18-602-383 12 kg Poly Pail
18-602-384 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-380 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1058500GMCS6
Add to Cart
Add to Cart

E1058, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Edetate Disodium, Dihydrate, BiotechGrade is an aminopolycarboxylic acid and a colorless, water-soluble solid. It is useful in the laboratory as a hexadentate ligand and chelating agent. The GR brand is an EMD Chemical trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 8.7-11.4%
Packaging Poly Bottle
Quantity 6 x 500 g
Percent Purity 99 to 101%
Grade Biotech
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.