Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287326
Description
EDF1 Polyclonal specifically detects EDF1 in Human samples. It is validated for Western Blot.Specifications
EDF1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Protein A purified | |
RUO | |
8721 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
EDF-1Multiprotein-bridging factor 1, endothelial differentiation-related factor 1, MBF1, MGC9058, multiprotein bridging factor 1, multiprotein bridging factor-1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human EDF1 | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction