Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDF1 Monoclonal Antibody (OTI7B6)
GREENER_CHOICE

Catalog No. PIMA527354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527354 Supplier Invitrogen™ Supplier No. MA527354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EDF1 Monoclonal Antibody for Western Blot

This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDF1
Applications Western Blot
Classification Monoclonal
Clone OTI7B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EDF1
Gene Accession No. O60869
Gene Alias 0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829
Gene Symbols EDF1
Host Species Mouse
Immunogen Full length human recombinant protein of EDF1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8721
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.