Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596104 Supplier Invitrogen™ Supplier No. PA596104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAESDWDTVT VLRKKGPTAA QAKSKQAILA AQRRGEDVET SKKWAAGQNK QHSITKNTAK LDRETEELHH DRVTLEVGKV IQQGRQSKGL TQKDLATKIN EKPQVIADYE SGRAIPNNQV LGKIERAIGL KLRGKDIGKP IEKGPRAK; Positive Samples: HepG2, Mouse brain, Mouse liver, Rat brain; Cellular Location: Cytoplasm, Nucleus.

This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EDF1
Gene Accession No. O60869, P69736, Q9JMG1
Gene Alias 0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829
Gene Symbols EDF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human EDF1 (NP_0037831).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296570, 59022, 8721
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.