Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106830
Description
Antibody detects endogenous levels of total EDG2.
EDG2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis.
Specifications
EDG2 | |
Polyclonal | |
Unconjugated | |
LPAR1 | |
AI326300; Edg2; edg-2; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; Gpcr26; Gpcr91; GPR26; G-protein coupled receptor 26; Kdt2; LPA receptor 1; LPA1; LPA-1; LPAR1; Lysophosphatidic acid receptor 1; lysophosphatidic acid receptor Edg-2; Mrec1.3; rec.1.3; rec1.3; ventricular zone gene 1; Vzg1; vzg-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116744, 14745, 1902 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61793, P61794, Q92633 | |
LPAR1 | |
A synthesized peptide derived from human LPAR1(Accession Q92633), corresponding to amino acid residues I11-I61. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction