Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG2 Recombinant Rabbit Monoclonal Antibody (3U1Q0)
GREENER_CHOICE

Catalog No. PIMA542724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542724 Supplier Invitrogen™ Supplier No. MA542724
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HeLa, Jurkat, Mouse lung, Mouse brain, Rat brain. Immunogen sequence: MAAISTSIPV ISQPQFTAMN EPQCFYNESI AFFYNRSGKH LATEWNTVSK LVMGLGITVC IFIMLANLLV MVAIYVNRRF HFPIYYLMAN LAAADFFAGL.

EDG2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3U1Q0
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPAR1
Gene Accession No. P61793, P61794, Q92633
Gene Alias AI326300; Edg2; edg-2; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; Gpcr26; Gpcr91; GPR26; G-protein coupled receptor 26; Kdt2; LPA receptor 1; LPA1; LPA-1; LPAR1; Lysophosphatidic acid receptor 1; lysophosphatidic acid receptor Edg-2; Mrec1.3; rec.1.3; rec1.3; ventricular zone gene 1; Vzg1; vzg-1
Gene Symbols LPAR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EDG2/LPA-1 (Q92633).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116744, 14745, 1902
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.