Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDG3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142474
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
EDG-3 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG-3 is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG-3 is expressed in cardiovascular, leukocyte-containing and other tissues.
Specifications
| EDG3 | |
| Polyclonal | |
| Unconjugated | |
| S1PR3 | |
| AI132464; cardiac sphingosine-1-phosphate specific receptor; EDG3; EDG-3; Edg3l; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; FLJ37523; FLJ93220; G protein-coupled receptor, endothelial differentiation gene-3; Lpb3; Lysophospholipid receptor B3; MGC71696; S1P receptor 3; S1P receptor EDG3; S1P receptor Edg-3; S1P receptor S1P3; S1p3; S1PR3; sphingosine; Sphingosine 1-phosphate receptor 3; sphingosine 1-phosphate receptor Edg-3; sphingosine-1-phosphate receptor 3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1903 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q99500 | |
| S1PR3 | |
| Peptide with sequence C-SSRKVANHNNSERS (aa 225-238). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction