Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513129
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513129 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513129 Supplier Invitrogen™ Supplier No. PA513129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EDG3 Polyclonal Antibody for Western Blot

EDG-3 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG-3 is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG-3 is expressed in cardiovascular, leukocyte-containing and other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG3
Applications Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene S1PR3
Gene Accession No. Q99500, Q9Z0U9
Gene Alias AI132464; cardiac sphingosine-1-phosphate specific receptor; EDG3; EDG-3; Edg3l; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; FLJ37523; FLJ93220; G protein-coupled receptor, endothelial differentiation gene-3; Lpb3; Lysophospholipid receptor B3; MGC71696; S1P receptor 3; S1P receptor EDG3; S1P receptor Edg-3; S1P receptor S1P3; S1p3; S1PR3; sphingosine; Sphingosine 1-phosphate receptor 3; sphingosine 1-phosphate receptor Edg-3; sphingosine-1-phosphate receptor 3
Gene Symbols S1PR3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human EDG3.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13610, 1903
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.