Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG3 Recombinant Rabbit Monoclonal Antibody (5E3G9)
GREENER_CHOICE

Catalog No. PIMA537848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537848 Supplier Invitrogen™ Supplier No. MA537848
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, PC-3, Mouse lung, Mouse kidney, Mouse thymus, Rat lung, Rat spleen Immunogen sequence: MATALPPRLQ PVRGNETLRE HYQYVGKLAG RLKEASEGST LTTVLFLVIC SFIVLENLMV LIAIWKNNKF HNRMYFFIGN LALCDLLAGI AYKVNILMSG.

EDG-3 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG-3 is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG-3 is expressed in cardiovascular, leukocyte-containing and other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5E3G9
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene S1PR3
Gene Accession No. Q99500, Q9Z0U9
Gene Alias AI132464; cardiac sphingosine-1-phosphate specific receptor; EDG3; EDG-3; Edg3l; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; FLJ37523; FLJ93220; G protein-coupled receptor, endothelial differentiation gene-3; Lpb3; Lysophospholipid receptor B3; MGC71696; S1P receptor 3; S1P receptor EDG3; S1P receptor Edg-3; S1P receptor S1P3; S1p3; S1PR3; sphingosine; Sphingosine 1-phosphate receptor 3; sphingosine 1-phosphate receptor Edg-3; sphingosine-1-phosphate receptor 3
Gene Symbols S1PR3
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13610, 1903, 306792
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.