Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590061 Supplier Invitrogen™ Supplier No. PA590061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVVCWTPGQV VLLLDGLGCE SCNVLAVEKY FLLLAEANSL VNAAVYSCRD AEMRRTFRRL LCCACLRQST RESVHYTSSA QGGASTRIML PENGHPLMDS T; Positive Samples: HL-60; Cellular Location: Cell membrane, Cell surface, Multi-pass membrane protein.

Edg4 is a Lysophospholipid/Lysosphingolipid Receptor activated by phosphatidic acid (PA) and lysophosphatidic acid (LPA). It enhances the transcription of serum response element (SRE) reporter genes. In CD4(+) T cells, Edg4 may be involved in the control of interleukin-2 secretion. Edg4 expression has been documented in leukocytes, pancreas, spleen, thymus, prostate, and testis. Edg4 expression has also been seen in many cancer cell lines, including those of brain, blood, breast, ovary, cervix, colon, skin, and thyroid. ESTs have been isolated from B-cell/lung/testis, blood, brain, embryo, heart/melanocyte/uterus, liver, placenta, prostate, and skin libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LPAR2
Gene Accession No. Q9HBW0
Gene Alias Edg4; EDG-4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 4; G protein-coupled receptor; IPA2; LPA receptor 2; LPA receptor EDG4; LPA2; LPA-2; LPA2 Receptor; LPAR2; Lysophosphatidic acid receptor 2; lysophosphatidic acid receptor EDG4; lysophosphatidic acid receptor Edg-4; Pbx4; pre-B-cell leukemia transcription factor 4; RGD1561336
Gene Symbols LPAR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human LPAR2 (NP_0047112).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9170
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.