Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111621 Supplier Invitrogen™ Supplier No. PA5111621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSREVCRAVL SFLCCGCLRL GMRGPGDCLA RAVEAHSGAS TTDSSLRPRD S Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.

EDG6 (S1P4) belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG6 (S1P4) is sphingosine-1-phosphate. There are 8 known members of the EDG receptor family and they are implicated in mediating growth related effects such as induction of cellular proliferation, alterations in differentiation and survival and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C and several protein tyrosine kinases. EDG6 (S1P4) is expressed in lymphoid and hematopoietic tissue and in the lungs. Recently, the designation of the EDG receptors has been modified to include the ligand for the receptor. Thus, the new designation for the EDG6 (S1P4) receptor is the sphingosine 1-phosphate receptor-4 (S1P4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene S1PR4
Gene Accession No. O95977
Gene Alias Edg6; EDG-6; Endothelial differentiation G-protein coupled receptor 6; endothelial differentiation G-protein-coupled receptor 6; endothelial differentiation, G protein coupled receptor 6; endothelial differentiation, G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 6; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 6; LPC1; Lysophospholipid receptor C1; S1P receptor 4; S1P receptor Edg-6; S1P(4) receptor; S1p4; S1PR4; SLP4; sphingosine 1-phosphate receptor 4; Sphingosine 1-phosphate receptor Edg-6; sphingosine-1-phosphate receptor 4
Gene Symbols S1PR4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EDG6. Recombinant protein control fragment (Product #RP-105421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8698
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.