Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EDG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534185
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534185 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534185 Supplier Invitrogen™ Supplier No. PA534185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Marmoset (89%) Mouse, Rat, Hamster, Dog, Bat (83%).

The lysosphingolipid sphingosine 1-phosphate (S1P) regulates cell proliferation, apoptosis, motility, and neurite retraction. Its actions may be both intracellular as a second messenger and extracellular as a receptor ligand. S1P and the structurally related lysolipid mediator lysophosphatidic acid (LPA) signal cells through a set of G protein-coupled receptors known as EDG receptors. Some EDG receptors (e.g., EDG1) are S1P receptors; others (e.g., EDG2) are LPA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDG8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene S1PR5
Gene Accession No. Q9H228
Gene Alias Edg8; Edg-8; endothelial differentiation G-protein-coupled receptor 8; endothelial differentiation, sphingolipid G-protein-coupled receptor, 8; lpB4; Lysophospholipid receptor B4; nerve growth factor-regulated G-protein-coupled receptor 1; Nrg1; NRG-1; S1P receptor 5; S1P receptor Edg-8; S1P(5) receptor; S1P5; S1PR5; Sphingosine 1-phosphate receptor 5; sphingosine 1-phosphate receptor EDG8; sphingosine 1-phosphate receptor Edg-8; sphingosine-1-phosphate receptor 5; sphingosine-1-phosphate receptor LPB4; SPPR-1; SPPR-2
Gene Symbols S1PR5
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd cytoplasmic domain of human S1PR5 / EDG8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.